Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCTTTTAAAAAGTAAGCCATCCC[G/T]AGTGGATAGCCTATTTCATCAGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605623 MIM: 602632 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MKLN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MKLN1 - muskelin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PODXL - podocalyxin like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018111.2 | 3708 | UTR 3 | NP_001018121.1 | |||
NM_005397.3 | 3708 | UTR 3 | NP_005388.2 |