Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCTTCGTGCTTTTCCTCAAGCAG[A/G]TAACTAGGAGTATCCCTACTCCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616303 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
WDR91 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
WDR91 - WD repeat domain 91 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014149.3 | 3669 | UTR 3 | NP_054868.3 | |||
XM_011516083.2 | 3669 | UTR 3 | XP_011514385.1 | |||
XM_011516086.2 | 3669 | UTR 3 | XP_011514388.1 | |||
XM_017012070.1 | 3669 | UTR 3 | XP_016867559.1 | |||
XM_017012071.1 | 3669 | UTR 3 | XP_016867560.1 | |||
XM_017012072.1 | 3669 | UTR 3 | XP_016867561.1 |