Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAATTTGGTCTGCATTACCCAATA[C/T]GTTATATACAGCCGTCTTTTTATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606722 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NCALD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NCALD - neurocalcin delta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040624.1 | 1696 | UTR 3 | NP_001035714.1 | |||
NM_001040625.1 | 1696 | UTR 3 | NP_001035715.1 | |||
NM_001040626.1 | 1696 | UTR 3 | NP_001035716.1 | |||
NM_001040627.1 | 1696 | UTR 3 | NP_001035717.1 | |||
NM_001040628.1 | 1696 | UTR 3 | NP_001035718.1 | |||
NM_001040629.1 | 1696 | UTR 3 | NP_001035719.1 | |||
NM_001040630.1 | 1696 | UTR 3 | NP_001035720.1 | |||
NM_032041.2 | 1696 | UTR 3 | NP_114430.2 | |||
XM_011517332.2 | 1696 | UTR 3 | XP_011515634.1 | |||
XM_011517333.2 | 1696 | UTR 3 | XP_011515635.1 | |||
XM_011517334.2 | 1696 | UTR 3 | XP_011515636.1 | |||
XM_011517335.2 | 1696 | UTR 3 | XP_011515637.1 | |||
XM_017013900.1 | 1696 | Intron | XP_016869389.1 | |||
XM_017013901.1 | 1696 | UTR 3 | XP_016869390.1 | |||
XM_017013902.1 | 1696 | UTR 3 | XP_016869391.1 |