Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTGAGTTAGTCTATACCCCTACC[A/G]GAATGATCTCTGAAAATCAAATCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 100880 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACO1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACO1 - aconitase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278352.1 | Intron | NP_001265281.1 | ||||
NM_002197.2 | Intron | NP_002188.1 | ||||
XM_005251476.1 | Intron | XP_005251533.1 | ||||
XM_011517888.1 | Intron | XP_011516190.1 |