Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGAGAGCACTACGTCAGCAAATG[G/C]GGGGAGCAGTATTTCCCTTGGTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 137060 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
B4GALT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
B4GALT1 - beta-1,4-galactosyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001497.3 | 3565 | UTR 3 | NP_001488.2 | |||
XM_005251440.4 | 3565 | UTR 3 | XP_005251497.1 |