Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAAGAACCCCAGAATAATGCCCAC[A/T]GGGACAAAGAAGGTAAGTGGTCATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603179 MIM: 190990 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGEF39 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGEF39 - Rho guanine nucleotide exchange factor 39 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CA9 - carbonic anhydrase 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001216.2 | 433 | Missense Mutation | AGG,TGG | R,W 131 | NP_001207.2 |
TPM2 - tropomyosin 2 (beta) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |