Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACTTGCAAACAGTGCCGTCCAAT[A/G]TGAAAACCTTAAACAAACACTTCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608704 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NMRK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NMRK1 - nicotinamide riboside kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127603.1 | Intron | NP_001121075.1 | ||||
NM_017881.2 | Intron | NP_060351.1 | ||||
XM_006717162.1 | Intron | XP_006717225.1 | ||||
XM_006717163.2 | Intron | XP_006717226.1 | ||||
XM_011518817.1 | Intron | XP_011517119.1 | ||||
XM_011518818.1 | Intron | XP_011517120.1 | ||||
XM_011518819.2 | Intron | XP_011517121.1 | ||||
XM_017014868.1 | Intron | XP_016870357.1 | ||||
XM_017014869.1 | Intron | XP_016870358.1 | ||||
XM_017014870.1 | Intron | XP_016870359.1 | ||||
XM_017014871.1 | Intron | XP_016870360.1 | ||||
XM_017014872.1 | Intron | XP_016870361.1 | ||||
XM_017014873.1 | Intron | XP_016870362.1 | ||||
XM_017014874.1 | Intron | XP_016870363.1 |