Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCCAAAAAGCGCTTTGAATACTG[C/T]TCTGGGTTCACGGTGGAGATCTCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603140 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIP4K2A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PIP4K2A - phosphatidylinositol-5-phosphate 4-kinase type 2 alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005028.4 | 1063 | Silent Mutation | GAA,GAG | E,E 391 | NP_005019.2 | |
XM_006717450.2 | 1063 | Silent Mutation | GAA,GAG | E,E 340 | XP_006717513.1 | |
XM_011519496.1 | 1063 | Silent Mutation | GAA,GAG | E,E 332 | XP_011517798.1 | |
XM_017016330.1 | 1063 | Silent Mutation | GAA,GAG | E,E 332 | XP_016871819.1 | |
XM_017016331.1 | 1063 | Silent Mutation | GAA,GAG | E,E 332 | XP_016871820.1 | |
XM_017016332.1 | 1063 | Silent Mutation | GAA,GAG | E,E 281 | XP_016871821.1 |