Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGATAACTAGCCACCTGCAAGGGC[C/T]AAATCTCTCTGGGACGTTTGCACGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605319 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR3155A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR3155A - microRNA 3155a | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3155B - microRNA 3155b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PFKFB3 - 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145443.2 | Intron | NP_001138915.1 | ||||
NM_001282630.2 | Intron | NP_001269559.1 | ||||
NM_001314063.1 | Intron | NP_001300992.1 | ||||
NM_001323016.1 | Intron | NP_001309945.1 | ||||
NM_001323017.1 | Intron | NP_001309946.1 | ||||
NM_004566.3 | Intron | NP_004557.1 | ||||
XM_005252464.1 | Intron | XP_005252521.1 | ||||
XM_011519493.1 | Intron | XP_011517795.1 | ||||
XM_017016325.1 | Intron | XP_016871814.1 | ||||
XM_017016326.1 | Intron | XP_016871815.1 | ||||
XM_017016327.1 | Intron | XP_016871816.1 | ||||
XM_017016328.1 | Intron | XP_016871817.1 | ||||
XM_017016329.1 | Intron | XP_016871818.1 |