Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACATTTTACTTTTATTTTGTAGGA[C/T]TGAAAGCTAAACTTAGCTTTATACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612047 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
E2F8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
E2F8 - E2F transcription factor 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256371.1 | 3346 | UTR 3 | NP_001243300.1 | |||
NM_001256372.1 | 3346 | UTR 3 | NP_001243301.1 | |||
NM_024680.3 | 3346 | UTR 3 | NP_078956.2 | |||
XM_011520367.1 | 3346 | UTR 3 | XP_011518669.1 |