Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGGTCAAGCCCAGGGCCCCAGG[A/G]GCCCTTGAAGCATACCCGGGGTTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600623 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD82 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CD82 - CD82 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024844.1 | Intron | NP_001020015.1 | ||||
NM_002231.3 | Intron | NP_002222.1 | ||||
XM_006718223.2 | Intron | XP_006718286.1 | ||||
XM_011520067.2 | Intron | XP_011518369.1 |