Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTATAAGATGCAGACTGTATCATG[A/G]CACAGTGGGAAAGTCACCATGTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613184 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OR52I1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OR52I1 - olfactory receptor family 52 subfamily I member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRIM68 - tripartite motif containing 68 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304496.1 | 3148 | UTR 3 | NP_001291425.1 | |||
NM_018073.7 | 3148 | UTR 3 | NP_060543.5 |