Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCCTTAGCCTTTTTCTCAGGCAC[C/T]TCTGGCAAACCAAACTTCCTATATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614911 MIM: 613221 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1QTNF4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1QTNF4 - C1q and tumor necrosis factor related protein 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MTCH2 - mitochondrial carrier 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317231.1 | 1834 | Intron | NP_001304160.1 | |||
NM_001317232.1 | 1834 | Intron | NP_001304161.1 | |||
NM_001317233.1 | 1834 | Intron | NP_001304162.1 | |||
NM_014342.3 | 1834 | Intron | NP_055157.1 | |||
XM_006718172.2 | 1834 | Intron | XP_006718235.1 | |||
XM_011519959.2 | 1834 | UTR 3 | XP_011518261.1 | |||
XM_011519960.2 | 1834 | UTR 3 | XP_011518262.1 | |||
XM_011519961.2 | 1834 | UTR 3 | XP_011518263.1 | |||
XM_017017462.1 | 1834 | Intron | XP_016872951.1 | |||
XM_017017463.1 | 1834 | Intron | XP_016872952.1 |