Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCAGATGAGCTTGGCTCCATTATT[A/C]ATCAGCTTGAGCTGCCTGGTGGCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 186921 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LMO1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LMO1 - LIM domain only 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270428.1 | Intron | NP_001257357.1 | ||||
NM_002315.2 | Intron | NP_002306.1 | ||||
XM_006718228.3 | Intron | XP_006718291.1 | ||||
XM_011520098.2 | Intron | XP_011518400.1 | ||||
XM_011520099.2 | Intron | XP_011518401.1 | ||||
XM_011520100.1 | Intron | XP_011518402.1 |