Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAAATGAGCATTCTCTTCACCAT[C/T]GCACACTCAGCAAAATCTGATTCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602883 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLCO1A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLCO1A2 - solute carrier organic anion transporter family member 1A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021094.3 | 4122 | UTR 3 | NP_066580.1 | |||
NM_134431.3 | 4122 | UTR 3 | NP_602307.1 | |||
XM_005253474.3 | 4122 | UTR 3 | XP_005253531.1 | |||
XM_005253477.3 | 4122 | UTR 3 | XP_005253534.1 | |||
XM_011520818.1 | 4122 | UTR 3 | XP_011519120.1 | |||
XM_011520819.1 | 4122 | UTR 3 | XP_011519121.1 | |||
XM_011520820.2 | 4122 | UTR 3 | XP_011519122.1 | |||
XM_011520821.2 | 4122 | UTR 3 | XP_011519123.1 | |||
XM_017019849.1 | 4122 | Intron | XP_016875338.1 | |||
XM_017019850.1 | 4122 | Intron | XP_016875339.1 |