Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTATCACATCCAGTAATATGTTAAG[C/T]TCTGAAGATACGTTTTCATTTTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605600 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IPO8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IPO8 - importin 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190995.1 | 4721 | UTR 3 | NP_001177924.1 | |||
NM_006390.3 | 4721 | UTR 3 | NP_006381.2 | |||
XM_017018691.1 | 4721 | UTR 3 | XP_016874180.1 | |||
XM_017018692.1 | 4721 | UTR 3 | XP_016874181.1 | |||
XM_017018693.1 | 4721 | Intron | XP_016874182.1 |