Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACACATTTGCTAGTGACAAGAGC[C/T]GATATTCAAATCCAGATCTGTAACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615027 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM60A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM60A - family with sequence similarity 60 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135811.1 | Intron | NP_001129283.1 | ||||
NM_001135812.1 | Intron | NP_001129284.1 | ||||
NM_021238.2 | Intron | NP_067061.1 | ||||
XM_011520803.2 | Intron | XP_011519105.1 | ||||
XM_017019758.1 | Intron | XP_016875247.1 | ||||
XM_017019759.1 | Intron | XP_016875248.1 | ||||
XM_017019760.1 | Intron | XP_016875249.1 | ||||
XM_017019761.1 | Intron | XP_016875250.1 | ||||
XM_017019762.1 | Intron | XP_016875251.1 |