Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATTATGGAGTTCAGGTAACCCAC[A/G]GGCCATACACCAGTAACTAAGAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608771 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MED13L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MED13L - mediator complex subunit 13 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015335.4 | 7108 | UTR 3 | NP_056150.1 | |||
XM_011538080.2 | 7108 | UTR 3 | XP_011536382.1 | |||
XM_011538081.2 | 7108 | UTR 3 | XP_011536383.1 | |||
XM_011538082.2 | 7108 | UTR 3 | XP_011536384.1 | |||
XM_017019090.1 | 7108 | UTR 3 | XP_016874579.1 |