Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTGTCTATCAACTTCTTCCTTCT[C/T]TTTGCCTTTCATCTCCCTCTGCAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616711 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TAOK3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TAOK3 - TAO kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016281.3 | Intron | NP_057365.3 | ||||
XM_005253897.1 | Intron | XP_005253954.1 | ||||
XM_005253898.1 | Intron | XP_005253955.1 | ||||
XM_006719445.2 | Intron | XP_006719508.1 | ||||
XM_011538437.1 | Intron | XP_011536739.1 | ||||
XM_011538439.1 | Intron | XP_011536741.1 | ||||
XM_017019408.1 | Intron | XP_016874897.1 | ||||
XM_017019409.1 | Intron | XP_016874898.1 | ||||
XM_017019410.1 | Intron | XP_016874899.1 | ||||
XM_017019411.1 | Intron | XP_016874900.1 | ||||
XM_017019412.1 | Intron | XP_016874901.1 | ||||
XM_017019413.1 | Intron | XP_016874902.1 |