Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTTTACCGCTAAGGCTTGACCGCA[A/G]GCTTCCACAAATTGCCTGGAATGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607325 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DOCK9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DOCK9 - dedicator of cytokinesis 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130048.1 | 6214 | Silent Mutation | GCC,GCT | A,A 2025 | NP_001123520.1 | |
NM_001130049.1 | 6214 | Intron | NP_001123521.1 | |||
NM_001130050.1 | 6214 | Intron | NP_001123522.1 | |||
NM_001318849.1 | 6214 | Silent Mutation | GCC,GCT | A,A 2003 | NP_001305778.1 | |
NM_015296.2 | 6214 | Silent Mutation | GCC,GCT | A,A 2026 | NP_056111.1 | |
XM_005254034.3 | 6214 | Silent Mutation | GCC,GCT | A,A 2012 | XP_005254091.1 | |
XM_005254035.3 | 6214 | Silent Mutation | GCC,GCT | A,A 1981 | XP_005254092.1 | |
XM_006719922.1 | 6214 | Silent Mutation | GCC,GCT | A,A 2060 | XP_006719985.1 | |
XM_006719924.2 | 6214 | Silent Mutation | GCC,GCT | A,A 2049 | XP_006719987.1 | |
XM_006719925.1 | 6214 | Silent Mutation | GCC,GCT | A,A 2048 | XP_006719988.1 | |
XM_006719931.3 | 6214 | Silent Mutation | GCC,GCT | A,A 2026 | XP_006719994.1 | |
XM_006719932.1 | 6214 | Silent Mutation | GCC,GCT | A,A 2025 | XP_006719995.1 | |
XM_006719936.1 | 6214 | Silent Mutation | GCC,GCT | A,A 2014 | XP_006719999.1 | |
XM_006719937.3 | 6214 | Silent Mutation | GCC,GCT | A,A 2003 | XP_006720000.1 | |
XM_017020503.1 | 6214 | Silent Mutation | GCC,GCT | A,A 2059 | XP_016875992.1 | |
XM_017020504.1 | 6214 | Silent Mutation | GCC,GCT | A,A 2045 | XP_016875993.1 | |
XM_017020505.1 | 6214 | Silent Mutation | GCC,GCT | A,A 2036 | XP_016875994.1 | |
XM_017020506.1 | 6214 | Silent Mutation | GCC,GCT | A,A 2028 | XP_016875995.1 | |
XM_017020507.1 | 6214 | Silent Mutation | GCC,GCT | A,A 2022 | XP_016875996.1 | |
XM_017020508.1 | 6214 | Silent Mutation | GCC,GCT | A,A 2014 | XP_016875997.1 | |
XM_017020509.1 | 6214 | Silent Mutation | GCC,GCT | A,A 2046 | XP_016875998.1 | |
XM_017020510.1 | 6214 | Silent Mutation | GCC,GCT | A,A 2005 | XP_016875999.1 | |
XM_017020511.1 | 6214 | Silent Mutation | GCC,GCT | A,A 1999 | XP_016876000.1 | |
XM_017020512.1 | 6214 | Silent Mutation | GCC,GCT | A,A 2034 | XP_016876001.1 | |
XM_017020513.1 | 6214 | Silent Mutation | GCC,GCT | A,A 1991 | XP_016876002.1 | |
XM_017020514.1 | 6214 | Silent Mutation | GCC,GCT | A,A 2023 | XP_016876003.1 | |
XM_017020515.1 | 6214 | Silent Mutation | GCC,GCT | A,A 2017 | XP_016876004.1 | |
XM_017020516.1 | 6214 | Silent Mutation | GCC,GCT | A,A 2011 | XP_016876005.1 | |
XM_017020517.1 | 6214 | Silent Mutation | GCC,GCT | A,A 2004 | XP_016876006.1 | |
XM_017020518.1 | 6214 | Silent Mutation | GCC,GCT | A,A 2037 | XP_016876007.1 | |
XM_017020519.1 | 6214 | Silent Mutation | GCC,GCT | A,A 1992 | XP_016876008.1 | |
XM_017020520.1 | 6214 | Silent Mutation | GCC,GCT | A,A 1980 | XP_016876009.1 | |
XM_017020521.1 | 6214 | Silent Mutation | GCC,GCT | A,A 2014 | XP_016876010.1 |