Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAAGCAATGAATAACAGAAGATG[A/G]TGTAAAAAAGTCATATTTTTTCTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600519 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GTF2A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GTF2A2 - general transcription factor IIA subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320929.1 | 760 | UTR 3 | NP_001307858.1 | |||
NM_001320930.1 | 760 | UTR 3 | NP_001307859.1 | |||
NM_004492.2 | 760 | UTR 3 | NP_004483.1 |