Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGAAACACAGGTTCTTTGGAGTT[A/G]TAAGAATGCTGTATGTGTTCACTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603109 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC102723493 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC102723493 - uncharacterized LOC102723493 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SMAD3 - SMAD family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145102.1 | Intron | NP_001138574.1 | ||||
NM_001145103.1 | Intron | NP_001138575.1 | ||||
NM_001145104.1 | Intron | NP_001138576.1 | ||||
NM_005902.3 | Intron | NP_005893.1 | ||||
XM_011521559.2 | Intron | XP_011519861.1 | ||||
XM_011521560.1 | Intron | XP_011519862.1 |