Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATTTATTTTTGAAAAAAAAAAAA[A/G]GCACTATAGAGGCAAGACTATTATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602520 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP2K5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAP2K5 - mitogen-activated protein kinase kinase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206804.1 | Intron | NP_001193733.1 | ||||
NM_002757.3 | Intron | NP_002748.1 | ||||
NM_145160.2 | Intron | NP_660143.1 | ||||
XM_011521787.2 | Intron | XP_011520089.1 | ||||
XM_011521788.2 | Intron | XP_011520090.1 | ||||
XM_017022414.1 | Intron | XP_016877903.1 |