Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGAGGATTAAATAATGAAAAGGTA[A/G]TGCATATAAACATTACTTACAATAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609096 MIM: 612501 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXO22 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FBXO22 - F-box protein 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012170.3 | Intron | NP_036302.1 | ||||
NM_147188.2 | Intron | NP_671717.1 | ||||
XM_011521447.2 | Intron | XP_011519749.1 |
UBE2Q2 - ubiquitin conjugating enzyme E2 Q2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |