Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACACAAACATGAACACCCACAGG[A/T]ACAGCCGGTCCACCACCATAGCCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 118503 MIM: 118509 | ||||||||||||||||||||
Literature Links: |
CHRNA3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHRNA3 - cholinergic receptor nicotinic alpha 3 subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CHRNB4 - cholinergic receptor nicotinic beta 4 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000750.3 | 1245 | Missense Mutation | TAC,TTC | Y,F 462 | NP_000741.1 | |
NM_001256567.1 | 1245 | Missense Mutation | ACC,TCC | T,S 136 | NP_001243496.1 | |
XM_011521186.2 | 1245 | Missense Mutation | TAC,TTC | Y,F 459 | XP_011519488.1 | |
XM_011521187.2 | 1245 | Missense Mutation | TAC,TTC | Y,F 459 | XP_011519489.1 | |
XM_011521190.2 | 1245 | Missense Mutation | TAC,TTC | Y,F 388 | XP_011519492.1 | |
XM_011521191.2 | 1245 | Missense Mutation | TAC,TTC | Y,F 388 | XP_011519493.1 | |
XM_011521192.2 | 1245 | Missense Mutation | TAC,TTC | Y,F 280 | XP_011519494.1 | |
XM_017021885.1 | 1245 | Missense Mutation | TAC,TTC | Y,F 432 | XP_016877374.1 | |
XM_017021886.1 | 1245 | Intron | XP_016877375.1 | |||
XM_017021887.1 | 1245 | Intron | XP_016877376.1 | |||
XM_017021888.1 | 1245 | Intron | XP_016877377.1 | |||
XM_017021889.1 | 1245 | Intron | XP_016877378.1 |