Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCAATGGTTACCACTATCGTTTTC[A/C]ACCAATATACAGATGTTCGCTCAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603795 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LITAF PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LITAF - lipopolysaccharide induced TNF factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136472.1 | 2127 | UTR 3 | NP_001129944.1 | |||
NM_001136473.1 | 2127 | UTR 3 | NP_001129945.1 | |||
NM_004862.3 | 2127 | UTR 3 | NP_004853.2 | |||
XM_006720982.2 | 2127 | UTR 3 | XP_006721045.1 | |||
XM_006720983.3 | 2127 | UTR 3 | XP_006721046.1 | |||
XM_006720984.3 | 2127 | UTR 3 | XP_006721047.1 | |||
XM_006720985.3 | 2127 | UTR 3 | XP_006721048.1 | |||
XM_011522754.2 | 2127 | UTR 3 | XP_011521056.1 | |||
XM_017023896.1 | 2127 | UTR 3 | XP_016879385.1 |