Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTCCTCCGTCCTCCCTCTAGACC[A/G]GAATGGAAACGACCTTCGGACCCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606692 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRAF7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRAF7 - TNF receptor associated factor 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032271.2 | 226 | Missense Mutation | AGA,GGA | R,G 29 | NP_115647.2 | |
XM_005255627.4 | 226 | Missense Mutation | AGA,GGA | R,G 29 | XP_005255684.1 | |
XM_011522700.1 | 226 | UTR 5 | XP_011521002.1 |