Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACCTATTGCTACCAGGGTGCCTA[A/C]AAGGATGCCCACTGCCGACAGCTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603118 | ||||||||||||||||||||
Literature Links: |
CDH16 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDH16 - cadherin 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204744.1 | 2456 | Missense Mutation | GTA,TTA | V,L 769 | NP_001191673.1 | |
NM_001204745.1 | 2456 | Missense Mutation | GTA,TTA | V,L 752 | NP_001191674.1 | |
NM_001204746.1 | 2456 | Missense Mutation | GTA,TTA | V,L 694 | NP_001191675.1 | |
NM_004062.3 | 2456 | Missense Mutation | GTA,TTA | V,L 791 | NP_004053.1 | |
XM_005255770.2 | 2456 | Missense Mutation | GTA,TTA | V,L 613 | XP_005255827.1 | |
XM_011522807.1 | 2456 | Missense Mutation | GTA,TTA | V,L 728 | XP_011521109.1 |