Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCTATATCTGGACTGATTTTTCC[A/G]TTTCCATTCCCCCTTGTTCAAGCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602027 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TERF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TERF2 - telomeric repeat binding factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005652.4 | Intron | NP_005643.2 | ||||
XM_005256121.3 | Intron | XP_005256178.1 | ||||
XM_005256122.3 | Intron | XP_005256179.1 | ||||
XM_005256123.3 | Intron | XP_005256180.1 | ||||
XM_005256124.3 | Intron | XP_005256181.1 |