Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTATAGCCCAGAGCCCCCCTCACT[G/T]TATGGGGCACTGGTGAGTACAGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606196 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IRX6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IRX6 - iroquois homeobox 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024335.2 | 824 | Silent Mutation | CTG,CTT | L,L 97 | NP_077311.2 | |
XM_005256137.2 | 824 | Silent Mutation | CTG,CTT | L,L 97 | XP_005256194.1 |