Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGGATAAACATCCGTGACTGCAC[C/T]TCATGATCCATCACGCACGACGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614245 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACSF3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACSF3 - acyl-CoA synthetase family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127214.3 | 6689 | Intron | NP_001120686.1 | |||
NM_001243279.2 | 6689 | Intron | NP_001230208.1 | |||
NM_001284316.1 | 6689 | Intron | NP_001271245.1 | |||
NM_174917.4 | 6689 | Intron | NP_777577.2 | |||
XM_005256293.2 | 6689 | Intron | XP_005256350.1 | |||
XM_011522942.1 | 6689 | Intron | XP_011521244.1 | |||
XM_011522943.1 | 6689 | Intron | XP_011521245.1 | |||
XM_017023018.1 | 6689 | Intron | XP_016878507.1 | |||
XM_017023019.1 | 6689 | Intron | XP_016878508.1 | |||
XM_017023020.1 | 6689 | UTR 3 | XP_016878509.1 | |||
XM_017023021.1 | 6689 | Intron | XP_016878510.1 | |||
XM_017023022.1 | 6689 | Intron | XP_016878511.1 |
LINC00304 - long intergenic non-protein coding RNA 304 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC400558 - uncharacterized LOC400558 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |