Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGGCTTAAAGGCCAGTGCTGTG[C/T]CTGAAAGACATTTTGAACAGGCAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC284009 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC284009 - uncharacterized LOC284009 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
METTL16 - methyltransferase like 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024086.3 | 4305 | UTR 3 | NP_076991.3 | |||
XM_005256801.2 | 4305 | UTR 3 | XP_005256858.1 | |||
XM_017025045.1 | 4305 | UTR 3 | XP_016880534.1 | |||
XM_017025046.1 | 4305 | UTR 3 | XP_016880535.1 |