Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGAGCTGCTGGGCCTTTCTGCCC[A/G]ATTTGTGTCTGTGCCGTGTTTTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609106 MIM: 606717 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXO39 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FBXO39 - F-box protein 39 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153230.2 | 230 | Missense Mutation | AAT,GAT | N,D 21 | NP_694962.1 | |
XM_011523697.1 | 230 | Missense Mutation | AAT,GAT | N,D 21 | XP_011521999.1 | |
XM_017024283.1 | 230 | Missense Mutation | AAT,GAT | N,D 21 | XP_016879772.1 |
XAF1 - XIAP associated factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |