Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTCAATGGTGCGTCCATTCACAG[C/T]TTTACTAGCCATGAAACTGAAACAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609498 | ||||||||||||||||||||
Literature Links: |
FBXO47 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FBXO47 - F-box protein 47 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008777.2 | 1303 | Missense Mutation | ACT,GCT | T,A 344 | NP_001008777.2 | |
XM_011524865.2 | 1303 | Missense Mutation | ACT,GCT | T,A 318 | XP_011523167.1 | |
XM_011524866.2 | 1303 | Missense Mutation | ACT,GCT | T,A 287 | XP_011523168.1 | |
XM_011524867.2 | 1303 | Missense Mutation | ACT,GCT | T,A 344 | XP_011523169.1 |