Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGCCTTGGGTTTATGCAGCGTCA[C/T]TTTTGGAGATGTTGAAGCACTGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609692 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
WIPF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
WIPF2 - WAS/WASL interacting protein family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_133264.4 | Intron | NP_573571.1 | ||||
XM_005257083.2 | Intron | XP_005257140.1 | ||||
XM_005257084.2 | Intron | XP_005257141.1 | ||||
XM_011524412.1 | Intron | XP_011522714.1 | ||||
XM_011524413.1 | Intron | XP_011522715.1 |