Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCGTTTAATTTTGGATTAGTGCA[C/T]TGCTTTAGGACTGGACAGATGAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC00469 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LINC00469 - long intergenic non-protein coding RNA 469 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC100134391 - uncharacterized LOC100134391 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278587.1 | 126 | Silent Mutation | CAC,CAT | H,H 25 | NP_001265516.1 | |
XM_017023987.1 | 126 | Silent Mutation | CAC,CAT | H,H 42 | XP_016879476.1 |