Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCATATGTCATGTTAAACTGCCT[C/T]GTATACTTCCTGTGCTGGCAAAAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616375 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UNK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UNK - unkempt family zinc finger | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080419.2 | Intron | NP_001073888.2 | ||||
XM_006722145.3 | Intron | XP_006722208.1 | ||||
XM_006722148.2 | Intron | XP_006722211.1 | ||||
XM_011525400.1 | Intron | XP_011523702.1 | ||||
XM_011525401.2 | Intron | XP_011523703.1 | ||||
XM_017025247.1 | Intron | XP_016880736.1 | ||||
XM_017025248.1 | Intron | XP_016880737.1 | ||||
XM_017025249.1 | Intron | XP_016880738.1 | ||||
XM_017025250.1 | Intron | XP_016880739.1 |