Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGCAAGGTGAAGCTGCCCTACCA[C/T]GCCACGCTGGGCATCCAGAGGTGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616997 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ENPP7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ENPP7 - ectonucleotide pyrophosphatase/phosphodiesterase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178543.4 | 367 | Silent Mutation | CAC,CAT | H,H 110 | NP_848638.3 | |
XM_011524737.1 | 367 | Silent Mutation | CAC,CAT | H,H 183 | XP_011523039.1 |