Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACATGGCCAACTTCCAGGCTTCA[C/T]GAAACTCACTGCCTTTTTGCACAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608587 MIM: 604260 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GHDC PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GHDC - GH3 domain containing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STAT5B - signal transducer and activator of transcription 5B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012448.3 | 2683 | UTR 3 | NP_036580.2 | |||
XM_005257626.4 | 2683 | Intron | XP_005257683.1 | |||
XM_017024977.1 | 2683 | UTR 3 | XP_016880466.1 |