Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGTGCCCGTGTGCTCCTGGCGCA[C/T]GGCGCAGACGTGGGCAGGGAGAATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615124 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD13B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKRD13B - ankyrin repeat domain 13B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152345.4 | 323 | Silent Mutation | CAC,CAT | H,H 70 | NP_689558.4 | |
XM_005257920.4 | 323 | Silent Mutation | CAC,CAT | H,H 70 | XP_005257977.1 | |
XM_011524319.2 | 323 | Silent Mutation | CAC,CAT | H,H 62 | XP_011522621.1 | |
XM_011524320.2 | 323 | Silent Mutation | CAC,CAT | H,H 70 | XP_011522622.1 | |
XM_011524321.2 | 323 | Silent Mutation | CAC,CAT | H,H 70 | XP_011522623.1 | |
XM_017024174.1 | 323 | UTR 5 | XP_016879663.1 | |||
XM_017024175.1 | 323 | UTR 5 | XP_016879664.1 | |||
XM_017024176.1 | 323 | UTR 5 | XP_016879665.1 |
CORO6 - coronin 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |