Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAGTGTGTGGGATGTCATCATCGT[A/T]AGGGGTTACCACAAGTGGATGCCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606779 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CORO6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CORO6 - coronin 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SSH2 - slingshot protein phosphatase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282129.1 | 7437 | UTR 3 | NP_001269058.1 | |||
NM_001282130.1 | 7437 | Intron | NP_001269059.1 | |||
NM_001282131.1 | 7437 | Intron | NP_001269060.1 | |||
NM_033389.3 | 7437 | UTR 3 | NP_203747.2 | |||
XM_005258058.3 | 7437 | UTR 3 | XP_005258115.1 | |||
XM_005258059.2 | 7437 | UTR 3 | XP_005258116.1 | |||
XM_005258060.3 | 7437 | UTR 3 | XP_005258117.1 | |||
XM_006722149.3 | 7437 | UTR 3 | XP_006722212.1 | |||
XM_011525402.2 | 7437 | UTR 3 | XP_011523704.1 | |||
XM_011525404.2 | 7437 | UTR 3 | XP_011523706.1 | |||
XM_011525405.2 | 7437 | UTR 3 | XP_011523707.1 | |||
XM_011525406.2 | 7437 | UTR 3 | XP_011523708.1 | |||
XM_011525407.1 | 7437 | UTR 3 | XP_011523709.1 | |||
XM_017025251.1 | 7437 | UTR 3 | XP_016880740.1 | |||
XM_017025252.1 | 7437 | UTR 3 | XP_016880741.1 | |||
XM_017025253.1 | 7437 | Intron | XP_016880742.1 | |||
XM_017025254.1 | 7437 | Intron | XP_016880743.1 | |||
XM_017025255.1 | 7437 | Intron | XP_016880744.1 |