Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTACTGAGGTTACTAACCAGCTAT[C/G]AGTATATTATTTTCGATTTGTGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600940 MIM: 609735 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LIG3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LIG3 - DNA ligase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002311.4 | 3509 | Intron | NP_002302.2 | |||
NM_013975.3 | 3509 | Intron | NP_039269.2 | |||
XM_005257970.3 | 3509 | Intron | XP_005258027.1 | |||
XM_006721896.3 | 3509 | Intron | XP_006721959.1 | |||
XM_017024624.1 | 3509 | Intron | XP_016880113.1 |
RAD51L3-RFFL - RAD51L3-RFFL readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RFFL - ring finger and FYVE-like domain containing E3 ubiquitin protein ligase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017368.1 | 3509 | UTR 3 | NP_001017368.1 |