Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTTCCACTAAAGCCTGGTCATCA[A/C]TGGGAGTTGAGGAGCTGGGGTTGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601392 MIM: 601394 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCL14 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCL14 - C-C motif chemokine ligand 14 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CCL15-CCL14 - CCL15-CCL14 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CCL16 - C-C motif chemokine ligand 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004590.3 | 436 | Silent Mutation | CAA,CAC | Q,H 120 | NP_004581.1 | |
XM_005258020.4 | 436 | Missense Mutation | CAA,CAC | Q,H 120 | XP_005258077.1 |
LOC101927339 - uncharacterized LOC101927339 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |