Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAATATCCCTCCTGTCCTCCACAT[A/G]GTTGTGCAGGGCCATGTGTGAAGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606486 MIM: 139312 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHMP1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHMP1B - charged multivesicular body protein 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020412.4 | 2391 | UTR 3 | NP_065145.2 |
GNAL - G protein subunit alpha L | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142339.2 | 2391 | Intron | NP_001135811.1 | |||
NM_001261443.1 | 2391 | Intron | NP_001248372.1 | |||
NM_001261444.1 | 2391 | Intron | NP_001248373.1 | |||
NM_182978.3 | 2391 | Intron | NP_892023.1 | |||
XM_006722324.2 | 2391 | Intron | XP_006722387.1 |