Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAGCTCCGAACAGGAGCTCTCTA[C/T]CCTCTCTCTATTACACTCCGGGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614806 MIM: 603474 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DMRTC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DMRTC2 - DMRT like family C2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6797 - microRNA 6797 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPS19 - ribosomal protein S19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001022.3 | 99 | UTR 5 | NP_001013.1 | |||
NM_001321483.1 | 99 | Intron | NP_001308412.1 | |||
NM_001321484.1 | 99 | Intron | NP_001308413.1 | |||
NM_001321485.1 | 99 | UTR 5 | NP_001308414.1 | |||
XM_017027113.1 | 99 | UTR 5 | XP_016882602.1 |