Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCAGCTGGAATCTGAACCCAGAA[A/G]TACCCAGCCTGCAGCATGTGCCATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC61 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC61 - coiled-coil domain containing 61 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267723.1 | 984 | Intron | NP_001254652.1 | |||
XM_005259191.3 | 984 | UTR 5 | XP_005259248.1 | |||
XM_005259192.4 | 984 | Intron | XP_005259249.1 | |||
XM_011527257.2 | 984 | Intron | XP_011525559.1 |