Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCGAGCAGGTGTGTCCGTGGCTT[G/T]TGGGGCCCTCGGAGCAGAAGAGTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF541 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF541 - zinc finger protein 541 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277075.1 | 3738 | Missense Mutation | CAA,CAC | Q,H 1147 | NP_001264004.1 | |
XM_005259311.4 | 3738 | Missense Mutation | CAA,CAC | Q,H 1147 | XP_005259368.1 | |
XM_005259314.3 | 3738 | Missense Mutation | CAA,CAC | Q,H 509 | XP_005259371.1 | |
XM_011527367.2 | 3738 | Missense Mutation | CAA,CAC | Q,H 1152 | XP_011525669.1 | |
XM_011527368.2 | 3738 | Missense Mutation | CAA,CAC | Q,H 1152 | XP_011525670.1 | |
XM_011527369.2 | 3738 | Missense Mutation | CAA,CAC | Q,H 1152 | XP_011525671.1 | |
XM_011527370.2 | 3738 | Missense Mutation | CAA,CAC | Q,H 1152 | XP_011525672.1 | |
XM_011527371.2 | 3738 | Missense Mutation | CAA,CAC | Q,H 1152 | XP_011525673.1 | |
XM_011527372.2 | 3738 | Intron | XP_011525674.1 | |||
XM_011527373.2 | 3738 | Missense Mutation | CAA,CAC | Q,H 1094 | XP_011525675.1 | |
XM_011527374.2 | 3738 | Missense Mutation | CAA,CAC | Q,H 1090 | XP_011525676.1 | |
XM_011527375.1 | 3738 | Intron | XP_011525677.1 | |||
XM_017027351.1 | 3738 | Missense Mutation | CAA,CAC | Q,H 1152 | XP_016882840.1 |