Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTCTGCTGGACCCGAGGCCTCCG[C/T]TCGAATCAGACGCTGTGCCTCCGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615038 MIM: 603996 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC114 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC114 - coiled-coil domain containing 114 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144577.3 | 1454 | Missense Mutation | AAC,AGC | N,S 639 | NP_653178.3 | |
XM_005259413.3 | 1454 | Missense Mutation | AAC,AGC | N,S 676 | XP_005259470.1 | |
XM_005259414.3 | 1454 | Missense Mutation | AAC,AGC | N,S 676 | XP_005259471.1 | |
XM_005259415.3 | 1454 | Missense Mutation | AAC,AGC | N,S 676 | XP_005259472.1 | |
XM_005259416.4 | 1454 | Missense Mutation | AAC,AGC | N,S 448 | XP_005259473.1 | |
XM_011527515.2 | 1454 | Missense Mutation | AAC,AGC | N,S 639 | XP_011525817.1 | |
XM_011527516.2 | 1454 | Missense Mutation | AAC,AGC | N,S 639 | XP_011525818.1 | |
XM_011527517.1 | 1454 | Intron | XP_011525819.1 | |||
XM_017027483.1 | 1454 | Missense Mutation | AAC,AGC | N,S 584 | XP_016882972.1 |
ZNF114 - zinc finger protein 114 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |