Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATGGTTTCAAATACGACAACGAC[A/G]ATGATGGGGAAGACTATCCAGTAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607972 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC6A16 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC6A16 - solute carrier family 6 member 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014037.2 | 1975 | Silent Mutation | ATC,ATT | I,I 575 | NP_054756.2 | |
XM_005258820.2 | 1975 | Silent Mutation | ATC,ATT | I,I 575 | XP_005258877.1 | |
XM_006723168.3 | 1975 | Silent Mutation | ATC,ATT | I,I 575 | XP_006723231.1 | |
XM_011526859.2 | 1975 | Silent Mutation | ATC,ATT | I,I 620 | XP_011525161.2 | |
XM_011526860.2 | 1975 | Silent Mutation | ATC,ATT | I,I 597 | XP_011525162.1 | |
XM_011526861.2 | 1975 | Silent Mutation | ATC,ATT | I,I 428 | XP_011525163.1 | |
XM_017026712.1 | 1975 | Silent Mutation | ATC,ATT | I,I 657 | XP_016882201.1 |