Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCACGTTCTCCACGCTCTCGGGGG[C/T]CACGTCGAACACCAGCGCCTCATTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600327 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SYT3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SYT3 - synaptotagmin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160328.1 | 1718 | Missense Mutation | NP_001153800.1 | |||
NM_001160329.1 | 1718 | Missense Mutation | NP_001153801.1 | |||
NM_032298.2 | 1718 | Missense Mutation | NP_115674.1 | |||
XM_011527390.2 | 1718 | Missense Mutation | XP_011525692.1 | |||
XM_011527391.2 | 1718 | Missense Mutation | XP_011525693.1 |